Ethyl 4-chloro-3-fluorobenzoate 95+%

Lieferant: Apollo Scientific

PC201192-25G
APOSPC201192-25GEA 81.2 EUR
APOSPC201192-25G
Ethyl 4-chloro-3-fluorobenzoate 95+%
Ethyl 4-chloro-3-fluorobenzoate
Formel: C₉H₈ClFO₂
Molecular Weight: 202,61 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06204445
CAS-Nummer: 203573-08-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR