3-(2,2,2-Trifluoroacetyl)benzoic acid 95+%

Lieferant: Apollo Scientific
Achtung

PC201194-100MG PC201194-250MG
APOSPC201194-100MGEA 470 EUR
APOSPC201194-100MG APOSPC201194-250MG
3-(2,2,2-Trifluoroacetyl)benzoic acid 95+%
3-(2,2,2-Trifluoroacetyl)benzoic acid
Formel: C₉H₅F₃O₃
Molecular Weight: 218.129 g/mol
Schmelzpunkt: 94…95 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD13173376
CAS-Nummer: 213598-05-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR