1-Chloro-6-fluoroisoquinoline 95+%

Lieferant: Apollo Scientific

PC201195-100MG PC201195-1G PC201195-250MG
APOSPC201195-100MGEA 101 EUR
APOSPC201195-100MG APOSPC201195-1G APOSPC201195-250MG
1-Chloro-6-fluoroisoquinoline 95+%
1-Chloro-6-fluoroisoquinoline
Formel: C₉H₅ClFN
Molecular Weight: 181.594 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09959717
CAS-Nummer: 214045-86-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR