5-(Trifluoromethyl)-1,2,3,4-tetrahydroisoquinoline hydrochloride 95%

Lieferant: Apollo Scientific
Achtung

PC201197-250MG
APOSPC201197-250MGEA 252 EUR
APOSPC201197-250MG
5-(Trifluoromethyl)-1,2,3,4-tetrahydroisoquinoline hydrochloride 95%
5-(Trifluoromethyl)-1,2,3,4-tetrahydroisoquinoline hydrochloride
Formel: C₁₀H₁₀F₃N·ClH
Molecular Weight: 237.649 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05861552
CAS-Nummer: 1074764-69-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR