3-Fluoro-N-methoxy-N-methylbenzamide 95+%

Lieferant: Apollo Scientific

PC201203-1G
APOSPC201203-1GEA 0 EUR
APOSPC201203-1G
3-Fluoro-N-methoxy-N-methylbenzamide 95+%
3-Fluoro-N-methoxy-N-methylbenzamide
Formel: C₉H₁₀FNO₂
Molecular Weight: 183,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02684305
CAS-Nummer: 226260-01-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR