7-Fluorophthalazin-1(2H)-one 97%
Lieferant: Apollo Scientific
PC201210-100MG
APOSPC201210-100MGEA
221
EUR
APOSPC201210-100MG
7-Fluorophthalazin-1(2H)-one 97%
7-Fluorophthalazin-1(2H)-one
Formel:
C₈H₅FN₂O Molecular Weight: 164.137 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28100454 CAS-Nummer: 23928-52-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...