2-Cyano-N-[4-(trifluoromethyl)phenyl]acetamide 97%
Lieferant: Apollo Scientific
PC201213-5G
APOSPC201213-5GEA
206
EUR
APOSPC201213-5G
2-Cyano-N-[4-(trifluoromethyl)phenyl]acetamide 97%
2-Cyano-N-[4-(trifluoromethyl)phenyl]acetamide
Formel:
C₁₀H₇F₃N₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD05669245 CAS-Nummer: 24522-30-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...