4,5-Difluorindol 95+%
Lieferant: Apollo Scientific
Synonyme:
4,5-Difluoro-1H-indole
PC201214-1G
APOSPC201214-1GEA
166
EUR
APOSPC201214-1G
4,5-Difluorindol 95+%
4,5-Difluoroindole
Formel:
C₈H₅F₂N Molecular Weight: 153,13 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01863154 CAS-Nummer: 247564-63-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...