6-Chloro-2-(trifluoromethyl)nicotinic acid

Lieferant: Apollo Scientific
Achtung

PC201216-1G
APOSPC201216-1GEA 328 EUR
APOSPC201216-1G
6-Chloro-2-(trifluoromethyl)nicotinic acid
6-Chloro-2-(trifluoromethyl)nicotinic acid
Formel: C₇H₃ClF₃NO₂
Schmelzpunkt: 160…162 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11840271
CAS-Nummer: 261635-83-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR