Methyl 4-cyano-3-fluorobenzoate 97%

Lieferant: Apollo Scientific

PC201220-5G
APOSPC201220-5GEA 92.8 EUR
APOSPC201220-5G
Methyl 4-cyano-3-fluorobenzoate 97%
Methyl 4-cyano-3-fluorobenzoate
Formel: C₉H₆FNO₂
Molecular Weight: 179,148 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09264509
CAS-Nummer: 268734-34-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR