4-[4-(Trifluormethyl)phenoxy]piperidin Hydrochlorid 95+%

Lieferant: Apollo Scientific

Synonyme: 4-(4-(Trifluoromethyl)phenoxy)piperidine hydrochloride

PC201221-250MG
APOSPC201221-250MGEA 52.2 EUR
APOSPC201221-250MG
4-[4-(Trifluormethyl)phenoxy]piperidin Hydrochlorid 95+%
4-[4-(Trifluormethyl)phenoxy]piperidin Hydrochlorid
Formel: C₁₂H₁₅ClF₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD02258937
CAS-Nummer: 287952-09-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR