3,5-Difluor-4-hydroxybenzonitril 96%

Lieferant: Apollo Scientific

PC201222-10G PC201222-5G
APOSPC201222-10GEA 303 EUR
APOSPC201222-10G APOSPC201222-5G
3,5-Difluor-4-hydroxybenzonitril 96%
3,5-Difluor-4-hydroxybenzonitril
Formel: C₇H₃F₂NO
Lagertemperatur: Raumtemperatur
MDL: MFCD09033181
CAS-Nummer: 2967-54-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR