3,5-Difluor-4-hydroxybenzonitril 96%
Lieferant: Apollo Scientific
PC201222-10G
PC201222-5G
APOSPC201222-10GEA
303
EUR
APOSPC201222-10G
APOSPC201222-5G
3,5-Difluor-4-hydroxybenzonitril 96%
3,5-Difluor-4-hydroxybenzonitril
Formel:
C₇H₃F₂NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD09033181 CAS-Nummer: 2967-54-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...