2-(3-Fluorophenyl)pyrrolidine 95+%

Lieferant: Apollo Scientific

PC201223-1G
APOSPC201223-1GEA 150 EUR
APOSPC201223-1G
2-(3-Fluorophenyl)pyrrolidine 95+%
2-(3-Fluorophenyl)pyrrolidine
Formel: C₁₀H₁₂FN
Molecular Weight: 165,2074 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02663475
CAS-Nummer: 298690-72-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR