2-Fluor-1-iod-4-nitrobenzol 95+%

Lieferant: Apollo Scientific
Achtung

PC201225-10G PC201225-5G PC201225-1G
APOSPC201225-10GEA 174 EUR
APOSPC201225-10G APOSPC201225-5G APOSPC201225-1G
2-Fluor-1-iod-4-nitrobenzol 95+%
2-Fluor-1-iod-4-nitrobenzol
Formel: C₆H₃FINO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00033987
CAS-Nummer: 2996-30-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR