7,8-Difluoroquinoline-3-carboxylic acid 95+%

Lieferant: Apollo Scientific

PC201229-1G
APOSPC201229-1GEA 238 EUR
APOSPC201229-1G
7,8-Difluoroquinoline-3-carboxylic acid 95+%
7,8-Difluoroquinoline-3-carboxylic acid
Formel: C₁₀H₅F₂NO₂
Molecular Weight: 209,149 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24483950
CAS-Nummer: 318685-41-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR