4-Fluor-1H-indazol 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Fluoroindazole
PC201232-1G
APOSPC201232-1GEA
21.9
EUR
APOSPC201232-1G
4-Fluor-1H-indazol 95+%
4-Fluoro-1H-indazole
Formel:
C₇H₅FN₂ Molecular Weight: 136,1264 g/mol Schmelzpunkt: 132 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07371561 CAS-Nummer: 341-23-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...