7-Fluor-1H-indazol 98%

Lieferant: Apollo Scientific
Achtung

PC201233-5G PC201233-1G
APOSPC201233-5GEA 43.5 EUR
APOSPC201233-5G APOSPC201233-1G
7-Fluor-1H-indazol 98%
7-Fluoro-1H-indazole
Formel: C₇H₅FN₂
Molecular Weight: 136.1264 g/mol
Schmelzpunkt: 123…127 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09026976
CAS-Nummer: 341-24-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR