5-(Benzyloxy)-2-fluoroaniline 95+%

Lieferant: Apollo Scientific

PC201234-1G PC201234-250MG PC201234-5G
APOSPC201234-1GEA 59.9 EUR
APOSPC201234-1G APOSPC201234-250MG APOSPC201234-5G
5-(Benzyloxy)-2-fluoroaniline 95+%
5-(Benzyloxy)-2-fluoroaniline
Formel: C₁₃H₁₂FNO
Lagertemperatur: Raumtemperatur
MDL: MFCD00835609
CAS-Nummer: 342042-82-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR