4-Fluorisatin 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Fluoroindoline-2,3-dione

PC201235-1G
APOSPC201235-1GEA 166 EUR
APOSPC201235-1G
4-Fluorisatin 95%
4-Fluorisatin
Formel: C₈H₄FNO₂
Molecular Weight: 165,1200 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01175824
CAS-Nummer: 346-34-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR