N,N-Dimethyl-4-fluorbenzolsulfonamid 95+%

Lieferant: Apollo Scientific

Synonyme: 4-Fluorbenzolsulfonsäuredimethylamid, N,N-Dimethyl-4-fluorbenzolsulfonamid

PC201245-5G
APOSPC201245-5GEA 132 EUR
APOSPC201245-5G
N,N-Dimethyl-4-fluorbenzolsulfonamid 95+%
N,N-Dimethyl-4-fluorbenzolsulfonamid
Formel: C₈H₁₀FNO₂S
Molecular Weight: 203,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01211990
CAS-Nummer: 383-31-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR