6-Chloro-3-(trifluoromethyl)picolinonitrile 95+%

Lieferant: Apollo Scientific

PC201249-250MG PC201249-1G PC201249-5G
APOSPC201249-250MGEA 81.2 EUR
APOSPC201249-250MG APOSPC201249-1G APOSPC201249-5G
6-Chloro-3-(trifluoromethyl)picolinonitrile 95+%
6-Chloro-3-(trifluoromethyl)picolinonitrile
Formel: C₇H₂ClF₃N₂
Molecular Weight: 206.552 g/mol
MDL: MFCD18261725
CAS-Nummer: 401590-41-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR