Methyl-6-fluorpicolinat 98%

Lieferant: Apollo Scientific

Synonyme: Methyl 6-fluoropicolinate

PC201257-1G
APOSPC201257-1GEA 120 EUR
APOSPC201257-1G
Methyl-6-fluorpicolinat 98%
Methyl-6-fluorpicolinat
Formel: C₇H₆FNO₂
Molecular Weight: 155,1300 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11110421
CAS-Nummer: 455-71-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR