6-Fluoro-1-naphthoic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC201264-250MG
APOSPC201264-250MGEA
127
EUR
APOSPC201264-250MG
6-Fluoro-1-naphthoic acid 97%
6-Fluoro-1-naphthoic acid
Formel:
C₁₁H₇FO₂ Schmelzpunkt: 234…236 °C Lagertemperatur: Raumtemperatur |
CAS-Nummer:
575-08-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...