6-Fluoro-1H-benzimidazole-2-thiol 95+%

Lieferant: Apollo Scientific

Synonyme: 5-Fluoro-1H-benzo[d]imidazole-2(3H)-thione

PC201265-5G
APOSPC201265-5GEA 127 EUR
APOSPC201265-5G
6-Fluoro-1H-benzimidazole-2-thiol 95+%
6-Fluoro-1H-benzimidazole-2-thiol
Formel: C₇H₅FN₂S
Molecular Weight: 168.19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07432806
CAS-Nummer: 583-42-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR