DL-β-(4-Fluorphenyl)alaninol 97%

Lieferant: Apollo Scientific

Synonyme: 3-Amino-3-(4-fluorophenyl)propan-1-ol

PC201269-1G
APOSPC201269-1GEA 84.1 EUR
APOSPC201269-1G
DL-β-(4-Fluorphenyl)alaninol 97%
DL-β-(4-Fluorphenyl)alaninol
Formel: C₉H₁₂FNO
Molecular Weight: 169,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03412718
CAS-Nummer: 612532-52-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR