2,4-Dichloro-3-fluoropyridine 97%

Lieferant: Apollo Scientific

PC201271-100MG PC201271-250MG PC201271-1G
APOSPC201271-100MGEA 82.9 EUR
APOSPC201271-100MG APOSPC201271-250MG APOSPC201271-1G
2,4-Dichloro-3-fluoropyridine 97%
2,4-Dichloro-3-fluoropyridine
Formel: C₅H₂Cl₂FN
Lagertemperatur: Raumtemperatur
MDL: MFCD12196994
CAS-Nummer: 628691-85-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR