Methyl 2-amino-3-(trifluoromethyl)benzoate 97%
Lieferant: Apollo Scientific
PC201275-1G
APOSPC201275-1GEA
85.7
EUR
APOSPC201275-1G
Methyl 2-amino-3-(trifluoromethyl)benzoate 97%
Methyl 2-amino-3-(trifluoromethyl)benzoate
Formel:
C₉H₈F₃NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11226293 CAS-Nummer: 64321-95-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...