5-Chloro-3-(difluoromethyl)-1-methyl-1H-pyrazole-4-carbaldehyde 98%
Lieferant: Apollo Scientific
PC201276-1G
APOSPC201276-1GEA
55.1
EUR
APOSPC201276-1G
5-Chloro-3-(difluoromethyl)-1-methyl-1H-pyrazole-4-carbaldehyde 98%
5-Chloro-3-(difluoromethyl)-1-methyl-1H-pyrazole-4-carbaldehyde
Formel:
C₆H₅ClF₂N₂O Molecular Weight: 194,567 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12827692 CAS-Nummer: 660845-30-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...