5-Bromo-8-fluoroisoquinoline 98%
Lieferant: Apollo Scientific
PC201279-1G
PC201279-5G
APOSPC201279-1GEA
317
EUR
APOSPC201279-1G
APOSPC201279-5G
5-Bromo-8-fluoroisoquinoline 98%
5-Bromo-8-fluoroisoquinoline
Formel:
C₉H₅BrFN Molecular Weight: 226.045 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18447885 CAS-Nummer: 679433-94-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...