6-(Trifluoromethyl)pyrazin-2-amine 97%

Lieferant: Apollo Scientific

PC201282-250MG
APOSPC201282-250MGEA 130 EUR
APOSPC201282-250MG
6-(Trifluoromethyl)pyrazin-2-amine 97%
6-(Trifluoromethyl)pyrazin-2-amine
Formel: C₅H₄F₃N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD10697803
CAS-Nummer: 69816-35-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR