2-Chloro-5-(trifluoromethyl)pyridin-3-amine 98%

Lieferant: Apollo Scientific
Achtung

PC201291-10G PC201291-5G PC201291-1G
APOSPC201291-10GEA 223 EUR
APOSPC201291-10G APOSPC201291-5G APOSPC201291-1G
2-Chloro-5-(trifluoromethyl)pyridin-3-amine 98%
2-Chloro-5-(trifluoromethyl)pyridin-3-amine
Formel: C₆H₄ClF₃N₂
Lagertemperatur: Kühlschrank
MDL: MFCD07375383
CAS-Nummer: 72587-18-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR