6-(2,2,2-Trifluoroethoxy)pyridin-3-amine 97%

Lieferant: Apollo Scientific

PC201293-250MG
APOSPC201293-250MGEA 82.9 EUR
APOSPC201293-250MG
6-(2,2,2-Trifluoroethoxy)pyridin-3-amine 97%
6-(2,2,2-Trifluoroethoxy)pyridin-3-amine
Formel: C₇H₇F₃N₂O
Molecular Weight: 192.138 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04111379
CAS-Nummer: 72617-82-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR