2-(Trifluoromethyl)pyrimidin-5-amine 98%

Lieferant: Apollo Scientific
Achtung

PC201294-250MG
APOSPC201294-250MGEA 68.3 EUR
APOSPC201294-250MG
2-(Trifluoromethyl)pyrimidin-5-amine 98%
2-(Trifluoromethyl)pyrimidin-5-amine
Formel: C₅H₄F₃N₃
Molecular Weight: 163,1000 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09754047
CAS-Nummer: 73418-87-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR