1-(4-Amino-3-fluorophenyl)ethanone 98%
Lieferant: Apollo Scientific
PC201298-5G
APOSPC201298-5GEA
77.1
EUR
APOSPC201298-5G
1-(4-Amino-3-fluorophenyl)ethanone 98%
1-(4-Amino-3-fluorophenyl)ethanone
Formel:
C₈H₈FNO Molecular Weight: 153,154 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12032124 CAS-Nummer: 73792-22-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...