3-Fluor-4-(trifluormethyl)benzylamin 98%

Lieferant: Apollo Scientific
Gefahr

PC2013-1G
APOSPC2013-1GEA 0 EUR
APOSPC2013-1G
3-Fluor-4-(trifluormethyl)benzylamin 98%
3-Fluor-4-(trifluormethyl)benzylamin
Formel: C₈H₇F₄N
Lagertemperatur: Raumtemperatur
MDL: MFCD00236287
CAS-Nummer: 235106-09-9
UN: 2735
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR