3-Ethynyl-4-fluoroaniline 95+%

Lieferant: Apollo Scientific

PC201303-5G
APOSPC201303-5GEA 640 EUR
APOSPC201303-5G
3-Ethynyl-4-fluoroaniline 95+%
3-Ethynyl-4-fluoroaniline
Formel: C8H6FN
Molecular Weight: 135,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09029299
CAS-Nummer: 77123-60-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR