5-Chloro-3-(trifluoromethyl)pyridin-2-amine 95+%
Lieferant: Apollo Scientific
PC201309-1G
APOSPC201309-1GEA
87.3
EUR
APOSPC201309-1G
5-Chloro-3-(trifluoromethyl)pyridin-2-amine 95+%
5-Chloro-3-(trifluoromethyl)pyridin-2-amine
Formel:
C₆H₄ClF₃N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD13185329 CAS-Nummer: 79456-33-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...