4-Ethyl-2-fluoroaniline 95+%
Lieferant: Apollo Scientific
PC201315-100MG
APOSPC201315-100MGEA
125
EUR
APOSPC201315-100MG
4-Ethyl-2-fluoroaniline 95+%
4-Ethyl-2-fluoroaniline
Formel:
C₈H₁₀FN Molecular Weight: 139,17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22480900 CAS-Nummer: 821791-69-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...