2-Amino-5-brom-6-fluorbenzonitril 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Amino-3-bromo-2-fluorobenzonitrile

PC201321-5G PC201321-1G PC201321-25G
APOSPC201321-5GEA 43.5 EUR
APOSPC201321-5G APOSPC201321-1G APOSPC201321-25G
2-Amino-5-brom-6-fluorbenzonitril 95%
2-Amino-5-brom-6-fluorbenzonitril
Formel: C₇H₄BrFN₂
Molecular Weight: 215.0225 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06657979
CAS-Nummer: 845866-92-4
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR