1-(4-Bromo-5-fluoro-2-hydroxyphenyl)ethanone 95+%

Lieferant: Apollo Scientific

PC201330-250MG
APOSPC201330-250MGEA 42.2 EUR
APOSPC201330-250MG
1-(4-Bromo-5-fluoro-2-hydroxyphenyl)ethanone 95+%
1-(4-Bromo-5-fluoro-2-hydroxyphenyl)ethanone
Formel: C₈H₆BrFO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11847403
CAS-Nummer: 865449-63-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR