cis-3,4-Difluoropyrrolidine hydrochloride 95+%

Lieferant: Apollo Scientific

PC201334-1G
APOSPC201334-1GEA 343 EUR
APOSPC201334-1G
cis-3,4-Difluoropyrrolidine hydrochloride 95+%
cis-3,4-Difluoropyrrolidine hydrochloride
Formel: C₄H₈ClF₂N
Lagertemperatur: Raumtemperatur
MDL: MFCD20230545
CAS-Nummer: 869481-94-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR