2-Chloro-3-fluoroisonicotinonitrile 97%

Lieferant: Apollo Scientific

PC201336-250MG
APOSPC201336-250MGEA 66.7 EUR
APOSPC201336-250MG
2-Chloro-3-fluoroisonicotinonitrile 97%
2-Chloro-3-fluoroisonicotinonitrile
Formel: C₆H₂ClFN₂
Molecular Weight: 156.545 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18802658
CAS-Nummer: 870065-56-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR