2-(4-Bromo-2,5-difluorophenyl)acetic acid 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC201338-1G
APOSPC201338-1GEA
58
EUR
APOSPC201338-1G
2-(4-Bromo-2,5-difluorophenyl)acetic acid 98%
2-(4-Bromo-2,5-difluorophenyl)acetic acid
Formel:
C₈H₅BrF₂O₂ Molecular Weight: 251.025 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11847567 CAS-Nummer: 871035-64-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...