2-(4-Fluorophenyl)cyclopropanecarboxylic acid 95+%

Lieferant: Apollo Scientific

PC201343-1G
APOSPC201343-1GEA 91.5 EUR
APOSPC201343-1G
2-(4-Fluorophenyl)cyclopropanecarboxylic acid 95+%
2-(4-Fluorophenyl)cyclopropanecarboxylic acid
Formel: C₁₀H₉FO₂
Molecular Weight: 180,176 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06804481
CAS-Nummer: 879324-64-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR