5-Bromo-6-trifluoromethylpyridin-2-ylamine 95+%

Lieferant: Apollo Scientific

PC201346-5G
APOSPC201346-5GEA 84.1 EUR
APOSPC201346-5G
5-Bromo-6-trifluoromethylpyridin-2-ylamine 95+%
5-Bromo-6-trifluoromethylpyridin-2-ylamine
Formel: C₆H₄BrF₃N₂
Molecular Weight: 241.0086 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11848010
CAS-Nummer: 882500-21-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR