6-Chloro-2-fluoropyridin-3-ol 98%

Lieferant: Apollo Scientific

Synonyme: 6-Chloro-2-fluoro-3-hydroxypyridine

PC201347-250MG
APOSPC201347-250MGEA 62.8 EUR
APOSPC201347-250MG
6-Chloro-2-fluoropyridin-3-ol 98%
6-Chloro-2-fluoropyridin-3-ol
Formel: C₅H₃ClFNO
Molecular Weight: 147.535 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07437887
CAS-Nummer: 883107-68-4

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR