2-Brom-3-fluor-4-methylpyridin 97%

Lieferant: Apollo Scientific

Synonyme: 2-Brom-3-fluor-4-picolin

PC201348-5G
APOSPC201348-5GEA 101 EUR
APOSPC201348-5G
2-Brom-3-fluor-4-methylpyridin 97%
2-Bromo-3-fluoro-4-methylpyridine
Formel: C₆H₅BrFN
Molecular Weight: 190,01 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04112499
CAS-Nummer: 884494-37-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR