6-Chloro-3-fluoropicolinaldehyde 98%

Lieferant: Apollo Scientific

PC201351-5G
APOSPC201351-5GEA 130 EUR
APOSPC201351-5G
6-Chloro-3-fluoropicolinaldehyde 98%
6-Chloro-3-fluoropicolinaldehyde
Formel: C₆H₃ClFNO
Lagertemperatur: Raumtemperatur
MDL: MFCD04972390
CAS-Nummer: 884494-77-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR