4-Chloro-5-fluoro-2-methylpyrimidine 96%

Lieferant: Apollo Scientific

PC201366-250MG PC201366-1G PC201366-5G
APOSPC201366-250MGEA 69.6 EUR
APOSPC201366-250MG APOSPC201366-1G APOSPC201366-5G
4-Chloro-5-fluoro-2-methylpyrimidine 96%
4-Chloro-5-fluoro-2-methylpyrimidine
Formel: C₅H₄ClFN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09834964
CAS-Nummer: 898044-50-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR